Assay Details
| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053931.3 | NP_446383.3 | 4 - 5 | 650 | 109 | |
| GenBank mRNA | AB027143.1 | - | 4 - 5 | 650 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept5
Genomic Map