Assay Details
| Gene Symbol: Eif4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008335.1 | NP_001008336.1 | 2 - 3 | 96 | 117 | |
| XM_006248534.2 | XP_006248596.1 | 2 - 3 | 112 | 117 | ||
| XM_006248535.2 | XP_006248597.1 | 2 - 3 | 112 | 117 | ||
| GenBank mRNA | BC085859.1 | - | 2 - 3 | 96 | 7190333 | 117 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif4a2
Genomic Map