Assay Details
| Gene Symbol: Paip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014148.1 | NP_001014170.1 | 3 - 4 | 307 | 76 | |
| XM_008772049.1 | XP_008770271.1 | 1 - 2 | 300 | 76 | ||
| GenBank mRNA | BC079261.1 | - | 3 - 4 | 307 | 7133501 | 76 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079261.1, NM_001014148.1, XM_008772049.1 transcripts.
Set Membership: |
|

Paip2
Genomic Map