Assay Details
| Gene Symbol: LOC103689930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254461.2 | XP_006254523.1 | 23 - 24 | 3765 | 129 | |
| XM_006254462.2 | XP_006254524.1 | 23 - 24 | 3762 | 129 | ||
| XM_008771994.1 | XP_008770216.1 | 23 - 24 | 3819 | 129 | ||
| Gene Symbol: Trappc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106160.1 | NP_001099630.1 | 3519 | 129 | ||
| GenBank mRNA | FQ230517.1 | - | 1376 | 129 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103689930
Genomic Map