Assay Details
| Gene Symbol: Zfp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013141.1 | NP_001013159.1 | 1 - 2 | 157 | 72 | |
| GenBank mRNA | BC089850.1 | - | 1 - 2 | 157 | 7375328 | 72 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC089850.1, NM_001013141.1 transcripts.
Set Membership: |
|

Zfp35
Genomic Map