Assay Details
| Gene Symbol: Eif4ebp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202552.1 | NP_001189481.1 | 2 - 3 | 311 | 80 | |
| GenBank mRNA | BF543463.1 | - | 2 - 3 | 310 | 80 | |
| BM391907.1 | - | 1 - 2 | 80 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Eif4ebp3
Genomic Map