Assay Details
Gene Symbol: Pcdha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053941.1 | NP_446393.1 | 1 | 2320 | 67 | |
GenBank mRNA | AB113386.1 | - | 1 | 2320 | 67 | |
AF539749.1 | - | 1 | 1943 | 67 | ||
AY573977.1 | - | 1 | 2320 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
