Assay Details
| Gene Symbol: Srp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106157.2 | NP_001099627.1 | 2 - 3 | 130 | 76 | |
| GenBank mRNA | BC167043.1 | - | 2 - 3 | 130 | 7319792 | 76 |
| FQ233350.1 | - | 2 - 3 | 180 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Srp19
Genomic Map