Assay Details
| Gene Symbol: RGD1562608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134607.1 | NP_001128079.1 | 7 - 8 | 1376 | 89 | |
| XM_003751756.3 | XP_003751804.2 | 9 - 10 | 1776 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1562608
Genomic Map