Assay Details
| Gene Symbol: RGD1562608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134607.1 | NP_001128079.1 | 8 - 9 | 1486 | 79 | |
| XM_003751756.3 | XP_003751804.2 | 10 - 11 | 1886 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154445 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P155136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74810 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154445 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P155136 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154445 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P155136 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P155136 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154445 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10803539 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800597 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800596 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10803537 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800594 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10803540 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10803538 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10800592 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10803536 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800001222.rn.1 | TC1800000174.rn.1 | RTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22418960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362216 | - | - |

RGD1562608
Genomic Map