Assay Details
| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014147.1 | NP_001014169.2 | 5 - 6 | 580 | 73 | |
| GenBank mRNA | BC079332.1 | - | 5 - 6 | 580 | 7096678 | 73 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tpgs2
Genomic Map