Assay Details
| Gene Symbol: Hmgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013184.1 | NP_001013202.1 | 3 - 4 | 155 | 128 | |
| GenBank mRNA | BC086423.1 | - | 3 - 4 | 155 | 7302011 | 128 |
| FQ220876.1 | - | 3 - 4 | 193 | 128 | ||
| FQ222121.1 | - | 3 - 4 | 194 | 128 | ||
| FQ230156.1 | - | 3 - 4 | 158 | 128 | ||
| Gene Symbol: LOC100911295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751029.3 | XP_003751077.1 | 3 - 4 | 226 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086423.1,NM_001013184.1,XM_003751029.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hmgn1
Genomic Map