Assay Details
Gene Symbol: Odf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191992.1 | NP_001178921.1 | 2 - 3 | 314 | 61 | |
XM_006242237.1 | XP_006242299.1 | 2 - 3 | 423 | 61 | ||
XM_006242238.2 | XP_006242300.1 | 3 - 4 | 957 | 61 | ||
XM_008765776.1 | XP_008763998.1 | 2 - 3 | 697 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
