Assay Details
| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031666.1 | NP_113854.1 | 1 - 2 | 149 | 72 | |
| GenBank mRNA | AF375463.1 | - | 1 - 2 | 149 | 72 | |
| DQ437524.1 | - | 1 - 2 | 149 | 72 | ||
| U85513.1 | - | 1 - 2 | 281 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syt10
Genomic Map