Assay Details
| Gene Symbol: Rabl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242227.1 | XP_006242289.1 | 1 - 2 | 97 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006242227.1 transcripts.
Set Membership: |
|

Rabl2a
Genomic Map