Assay Details
| Gene Symbol: Rabl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013221.1 | NP_001013239.1 | 3 - 4 | 479 | 78 | |
| XM_006242225.2 | XP_006242287.1 | 3 - 4 | 560 | 78 | ||
| XM_006242226.2 | XP_006242288.1 | 3 - 4 | 561 | 78 | ||
| XM_006242227.1 | XP_006242289.1 | 3 - 4 | 281 | 78 | ||
| XM_006242228.2 | XP_006242290.1 | 3 - 4 | 516 | 78 | ||
| GenBank mRNA | BC083624.1 | - | 3 - 4 | 479 | 7134121 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|

Rabl2a
Genomic Map