Assay Details
| Gene Symbol: Adm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201426.1 | NP_958829.1 | 1 - 2 | 158 | 91 | |
| GenBank mRNA | AB121036.1 | - | 1 - 2 | 158 | 91 | |
| AB181297.1 | - | 1 - 2 | 167 | 91 | ||
| FQ234235.1 | - | 1 - 2 | 151 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB121036.1, AB181297.1, NM_201426.1 transcripts.
Set Membership: |
|

Adm2
Genomic Map