Assay Details
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085485.1 | NP_001078954.1 | 2 - 3 | 873 | 70 | |
GenBank mRNA | BC166588.1 | - | 2 - 3 | 771 | 7929890 | 70 |
CK479732.1 | - | 1 - 2 | 202 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
