Assay Details
| Gene Symbol: Dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108070.1 | NP_001101540.1 | 8 - 9 | 1266 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108070.1 transcripts.
Set Membership: |
|

Dos
Genomic Map