Assay Details
| Gene Symbol: Dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108070.1 | NP_001101540.1 | 10 - 11 | 1595 | 77 | |
| GenBank mRNA | BC169056.1 | - | 1 - 2 | 207 | 7932746 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Dos
Genomic Map