Assay Details
| Gene Symbol: Cactin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081447.1 | NP_001074916.1 | 6 - 7 | 1412 | 87 | |
| XM_008765124.1 | XP_008763346.1 | 6 - 7 | 2516 | 87 | ||
| XM_008765125.1 | XP_008763347.1 | 6 - 7 | 2516 | 87 | ||
| XM_008765126.1 | XP_008763348.1 | 6 - 7 | 1903 | 87 | ||
| XM_008765127.1 | XP_008763349.1 | 6 - 7 | 1587 | 87 | ||
| GenBank mRNA | AY934505.1 | - | 6 - 7 | 1412 | 87 | |
| BC166406.1 | - | 6 - 7 | 2042 | 9028620 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cactin
Genomic Map