Assay Details
| Gene Symbol: Gzmm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057183.1 | NP_476531.1 | 1 - 2 | 98 | 125 | |
| GenBank mRNA | DV216119.1 | - | 1 - 2 | 98 | 125 | |
| FQ233430.1 | - | 1 - 2 | 125 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gzmm
Genomic Map