Assay Details
| Gene Symbol: Polr2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109614.1 | NP_001103084.1 | 1 - 2 | 129 | 65 | |
| XM_006241009.2 | XP_006241071.1 | 1 - 2 | 258 | 65 | ||
| GenBank mRNA | BC158787.1 | - | 1 - 2 | 87 | 7304442 | 65 |
| FQ219797.1 | - | 1 - 2 | 61 | 65 | ||
| FQ220392.1 | - | 1 - 2 | 59 | 65 | ||
| FQ230290.1 | - | 1 - 2 | 64 | 65 | ||
| FQ234550.1 | - | 1 - 2 | 109 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr2e
Genomic Map