Assay Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139252.1 | NP_640345.1 | 2 - 3 | 291 | 96 | |
| XM_008765155.1 | XP_008763377.1 | 2 - 3 | 274 | 96 | ||
| GenBank mRNA | AF503611.1 | - | 2 - 3 | 291 | 96 | |
| BC062088.1 | - | 2 - 3 | 247 | 5621318 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765155.1 transcripts.
Set Membership: |
|

Ppap2c
Genomic Map