Assay Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139252.1 | NP_640345.1 | 3 - 4 | 569 | 86 | |
| XM_008765155.1 | XP_008763377.1 | 3 - 4 | 552 | 86 | ||
| GenBank mRNA | AF503611.1 | - | 3 - 4 | 569 | 86 | |
| BC062088.1 | - | 3 - 4 | 525 | 5621318 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppap2c
Genomic Map