Assay Details
| Gene Symbol: Slc35f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105950.1 | NP_001099420.1 | 12 - 13 | 1480 | 65 | |
| XM_006249671.2 | XP_006249733.1 | 12 - 13 | 1377 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc35f5
Genomic Map