Assay Details
| Gene Symbol: Ddx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006996.1 | NP_001006997.1 | 9 - 10 | 1569 | 66 | |
| GenBank mRNA | BC083919.1 | - | 9 - 10 | 1569 | 7135005 | 66 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ddx18
Genomic Map