Assay Details
| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769507.1 | XP_008767729.1 | 3 - 4 | 305 | 92 | |
| XM_008769508.1 | XP_008767730.1 | 4 - 5 | 569 | 92 | ||
More Information
Set Membership: |
|

Nckap5
Genomic Map