Assay Details
| Gene Symbol: Maged2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271109.1 | NP_001258038.1 | 2 - 3 | 114 | 61 | |
| NM_001271110.1 | NP_001258039.1 | 2 - 3 | 142 | 61 | ||
| NM_080479.2 | NP_536727.2 | 2 - 3 | 114 | 61 | ||
| XM_006237924.2 | XP_006237986.1 | 2 - 3 | 204 | 61 | ||
| GenBank mRNA | AY724507.1 | - | 2 - 3 | 127 | 61 | |
| BC086998.1 | - | 2 - 3 | 108 | 61 | ||
| BC107467.1 | - | 2 - 3 | 114 | 7459969 | 61 | |
| FM059132.1 | - | 2 - 3 | 142 | 61 | ||
| FQ229095.1 | - | 2 - 3 | 127 | 61 | ||
More Information
Set Membership: |
|

Maged2
Genomic Map