Assay Details
| Gene Symbol: Maged2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271109.1 | NP_001258038.1 | 1 - 2 | 48 | 69 | |
| NM_080479.2 | NP_536727.2 | 1 - 2 | 48 | 69 | ||
| GenBank mRNA | BC086998.1 | - | 1 - 2 | 42 | 69 | |
| BC107467.1 | - | 1 - 2 | 48 | 7459969 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107467.1, NM_001271109.1, NM_080479.2 transcripts.
Set Membership: |
|

Maged2
Genomic Map