Assay Details
| Gene Symbol: LOC100909655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752022.3 | XP_003752070.1 | 842 | 152 | ||
| XM_008773107.1 | XP_008771329.1 | 898 | 152 | |||
| XM_008773109.1 | XP_008771331.1 | 2 - 3 | 898 | 152 | ||
| Gene Symbol: RragB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053972.2 | NP_446424.1 | 3 - 4 | 743 | 152 | |
| XM_008773066.1 | XP_008771288.1 | 2 - 3 | 898 | 152 | ||
| GenBank mRNA | BC078760.1 | - | 3 - 4 | 281 | 7123902 | 152 |
| BC087698.1 | - | 3 - 4 | 743 | 7314028 | 152 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100909655
Genomic Map