Assay Details
| Gene Symbol: Pfkfb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271063.1 | NP_001257992.1 | 3 - 4 | 368 | 74 | |
| NM_001271064.1 | NP_001257993.1 | 2 - 3 | 242 | 74 | ||
| NM_012621.4 | NP_036753.4 | 3 - 4 | 491 | 74 | ||
| XM_006256790.2 | XP_006256852.1 | 2 - 3 | 2401 | 74 | ||
| XM_006256791.2 | XP_006256853.1 | 3 - 4 | 576 | 74 | ||
| XM_008773123.1 | XP_008771345.1 | 5 - 6 | 542 | 74 | ||
| XM_008773124.1 | XP_008771346.1 | 5 - 6 | 538 | 74 | ||
| XM_008773125.1 | XP_008771347.1 | 4 - 5 | 416 | 74 | ||
| GenBank mRNA | AY707861.1 | - | 3 - 4 | 319 | 74 | |
| GQ422136.1 | - | 2 - 3 | 222 | 74 | ||
| J04197.1 | - | 3 - 4 | 521 | 74 | ||
| X15579.1 | - | 3 - 4 | 348 | 74 | ||
| Y00702.1 | - | 3 - 4 | 491 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkfb1
Genomic Map