Assay Details
| Gene Symbol: Klf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773103.1 | XP_008771325.1 | 1 - 2 | 66 | 130 | |
| XM_008773104.1 | XP_008771326.1 | 1 - 2 | 77 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008773103.1, XM_008773104.1 transcripts.
Set Membership: |
|

Klf8
Genomic Map