Assay Details
| Gene Symbol: Klf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773103.1 | XP_008771325.1 | 4 - 5 | 823 | 145 | |
| XM_008773104.1 | XP_008771326.1 | 4 - 5 | 834 | 145 | ||
| XM_008773105.1 | XP_008771327.1 | 5 - 6 | 1008 | 145 | ||
| XM_008773106.1 | XP_008771328.1 | 5 - 6 | 1059 | 145 | ||
More Information
Set Membership: |
|

Klf8
Genomic Map