Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008363.1 | NP_001008364.1 | 2 - 3 | 299 | 114 | |
| GenBank mRNA | BC085928.1 | - | 2 - 3 | 299 | 7110398 | 114 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085928.1, NM_001008363.1 transcripts.
Set Membership: |
|

Zfand2a
Genomic Map