Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008363.1 | NP_001008364.1 | 4 - 5 | 537 | 62 | |
| GenBank mRNA | BC085928.1 | - | 4 - 5 | 537 | 7110398 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfand2a
Genomic Map