Assay Details
| Gene Symbol: Ptcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109665.1 | NP_001103135.1 | 2 - 3 | 530 | 67 | |
| XM_006248871.1 | XP_006248933.1 | 2 - 3 | 559 | 67 | ||
| GenBank mRNA | BC129116.1 | - | 2 - 3 | 468 | 7113830 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Ptcd1
Genomic Map