Assay Details
| Gene Symbol: Gpr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037295.1 | NP_001032372.1 | 1 - 2 | 199 | 94 | |
| GenBank mRNA | BC127447.1 | - | 1 - 2 | 127 | 8363428 | 94 |
| X61496.1 | - | 1 - 2 | 199 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC127447.1, NM_001037295.1, X61496.1 transcripts.
Set Membership: |
|

Gpr12
Genomic Map