Assay Details
| Gene Symbol: Tubgcp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107516.1 | NP_001100986.1 | 15 - 16 | 2258 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tubgcp5| Gene Symbol: Tubgcp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107516.1 | NP_001100986.1 | 15 - 16 | 2258 | 64 | |
Set Membership: |
|