Assay Details
| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126296.2 | NP_001119768.2 | 4 - 5 | 679 | 75 | |
| XM_006236464.2 | XP_006236526.1 | 4 - 5 | 698 | 75 | ||
| XM_006236465.2 | XP_006236527.1 | 4 - 5 | 698 | 75 | ||
| XM_006236466.2 | XP_006236528.1 | 4 - 5 | 698 | 75 | ||
| GenBank mRNA | BC161978.1 | - | 4 - 5 | 679 | 9029930 | 75 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tra2a
Genomic Map