Assay Details
Gene Symbol: Lrrc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109231.1 | NP_001102701.1 | 1 - 2 | 158 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109231.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
