Assay Details
| Gene Symbol: Paxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107844.1 | NP_001101314.1 | 19 - 20 | 3101 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of histone H3-K36 methylation
vasculogenesis
cellular response to DNA damage stimulus
biological_process
response to ionizing radiation
DNA damage response, signal transduction by p53 class mediator
positive regulation of protein ubiquitination
positive regulation of histone acetylation
endothelial cell migration
positive regulation of isotype switching
histone H3-K4 methylation
positive regulation of histone H3-K4 methylation
positive regulation of transcription initiation from RNA polymerase II promoter
adipose tissue development
chorion development
molecular_function
positive regulation of histone H3-K36 methylation
vasculogenesis
cellular response to DNA damage stimulus
biological_process
response to ionizing radiation
DNA damage response, signal transduction by p53 class mediator
positive regulation of protein ubiquitination
positive regulation of histone acetylation
endothelial cell migration
positive regulation of isotype switching
histone H3-K4 methylation
positive regulation of histone H3-K4 methylation
positive regulation of transcription initiation from RNA polymerase II promoter
adipose tissue development
chorion development
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P034907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25097 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034907 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034907 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P034907 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P294467 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68582 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91961 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10852708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852715 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852705 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852706 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852710 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852707 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10852694 | - | - |
| Rat Genome 230 2.0 Array | 1376598_at | - | - |
| Rat Genome 230 2.0 Array | 1376598_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376598_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376598_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400000366.rn.1 | TC0400000064.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400000362.rn.1 | TC0400000064.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400000367.rn.1 | TC0400000064.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376598_at | - | - |
| Rat Expression Set 230 | 1376598_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852694 | - | - |
| Rat Genome U34 Set | rc_AI179993_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21532031 | - | - |

Paxip1
Genomic Map