Assay Details
| Gene Symbol: LOC100910079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235966.2 | XP_006236028.1 | 2 - 3 | 221 | 77 | |
| XM_006235968.2 | XP_006236030.1 | 2 - 3 | 2284 | 77 | ||
| Gene Symbol: Actr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191860.1 | NP_001178789.1 | 165 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910079
Genomic Map