Assay Details
| Gene Symbol: Mum1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257301.2 | XP_006257363.1 | 2 - 3 | 358 | 129 | |
| XM_006257305.2 | XP_006257367.1 | 2 - 3 | 251 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257301.2, XM_006257305.2 transcripts.
Set Membership: |
|

Mum1l1
Genomic Map