Assay Details
| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760532.1 | XP_008758754.1 | 5 - 6 | 535 | 87 | |
| XM_008760533.1 | XP_008758755.1 | 5 - 6 | 552 | 87 | ||
| GenBank mRNA | CO392889.1 | - | 411 | 87 | ||
| EF055485.1 | - | 8 - 9 | 708 | 87 | ||
| EF055488.1 | - | 8 - 9 | 708 | 87 | ||
More Information
Set Membership: |
|

RGD1311558
Genomic Map