Assay Details
| Gene Symbol: Lzts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231547.2 | XP_006231609.1 | 1 - 2 | 1055 | 86 | |
| GenBank mRNA | BC083580.1 | - | 1 - 2 | 54 | 7129196 | 86 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC083580.1, XM_006231547.2 transcripts.
Set Membership: |
|

Lzts2
Genomic Map