Assay Details
| Gene Symbol: Lbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047108.1 | NP_001040573.1 | 1 - 2 | 334 | 107 | |
| GenBank mRNA | AB197923.1 | - | 1 - 2 | 334 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lbx1
Genomic Map