Assay Details
| Gene Symbol: Sfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047856.2 | NP_001041321.2 | 1 - 2 | 76 | 96 | |
| GenBank mRNA | FM118556.1 | - | 1 - 2 | 76 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001047856.2 transcripts.
This assay is in the 5'UTR region of the NM_001047856.2 transcripts.
Set Membership: |
|

Sfr1
Genomic Map