Assay Details
| Gene Symbol: Pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053735.1 | NP_446187.1 | 2 - 3 | 686 | 155 | |
| GenBank mRNA | AY029199.1 | - | 2 - 3 | 686 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pi4k2a
Genomic Map