Assay Details
| Gene Symbol: Pdlim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017365.2 | NP_059061.1 | 4 - 5 | 589 | 68 | |
| GenBank mRNA | BC072465.1 | - | 4 - 5 | 580 | 7100446 | 68 |
| U23769.1 | - | 4 - 5 | 589 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdlim1
Genomic Map